About this item
This premium quality line of containers consists of 60 mL, 90 mL, wide 90 mL and 120 mL sizes for leakproof transportation
Each polypropylene container is lot numbered and has clear graduations. The wide 90 mL and 120 mL containers feature a QuickTurn™ cap closure for easy application. Sterile containers have an orange cap and tamper tab patient label. The patient identification labels meet current JCAHO standards. Non-sterile containers have a yellow cap and a non-tabbed patient label. The standard 90 mL container is also available in an opaque polypropylene for discrete transport. A space saver dispenser box of 20, 90 mL containers is convenient for laboratories and Doctor's offices. Ideal for Operating Room use, both the 90 mL and 120 mL containers are packaged in a sterile, clear plastic peel pouch.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.