AC Sensor, Brady Worldwide®
AC sensor
CA12222-724
: Avantor
Lightweight and simple to use, the AC Sensor was selected by OSHA electrical technical equipment committee as the only instrument of its type for use by OSHA compliance officers to identify and cite elctrical hazards of 50 volts or more.
Device uses AAA battery.
- Description:AC sensor