Nylon Lockout Hasp, Brady Worldwide®
Catalog # CA89008-106
Supplier: Brady Worldwide
Specifications
- Color:Orange
- Description:Nylon Lockout Hasp
- Material:Nylon
- Size:7"H x 1.75"W
- Cat. no.:CA89008-106
- Supplier No.:99668
Specifications
About this item
Nonconductive lockout hasps offer exceptional electrical insulating characteristics, and are made of orange nylon material
Each hasp can withstand up to 1,000 pounds, yet weighs only 1 ounce. Hasps accommodate up to six locks and have 3/8" diameter lock holes. Hasps are sold 1 per package.
Frequently Bought Together