Electrical Lockout Toolbox Kit, Brady Worldwide®
Supplier: Avantor
Certificates
About this item
The electrical lockout toolbox provides all the basics to lock out breakers, electrical plugs, wall switches, and fuses.
Delivery: Kit components includes 1 Mini Cable Lockout, 120V Snap-On Breaker Lockouts (2), 120/277V Clamp-On Breaker Lockouts (2), 480/600V Clamp-On Breaker Lockouts (2), 2 Universal Multi-Pole Breaker Lockouts, 1 Oversized Breaker Lockout, 2 Cleats for 120/277V Clamp-On Lockout, 2 Cleats for 480/600V Clamp-On Lockout, 3-In-1 Plug Lockout (1), 1 Wall Switch Lockout, 1 Small Fuse Blockout, 1 Large Fuse Blockout and 1 Extra-Large Lockout Toolbox.
Frequently Bought Together