Orders LinkContactUsLinkComponent
Safe-T-Grip® Fuse Puller, Brady
Safe-T-Grip® Fuse Puller, Brady
Supplier:   Avantor
Safe-T-Grip® Fuse Puller, Brady
Supplier:  Avantor
Certificates
Safe-T-Grip® Fuse Puller, Brady
Safe-T-Grip® Fuse Puller, Brady
2 Options Available Below
Product Details & Documents
Remove or replace cartridge fuses with ease by using the Safe-T-Grip® fuse pullers.
Notched handles ensure a no-slip grip. Amperage ranges are clearly marked and both types exceed MIL specifications. These yellow fuse pullers are designed for use with 0 to 100 A, 250 and 600 V fuses.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • CA12222-728
    Small Safe-T-Grip® Fuse Puller
    Small Safe-T-Grip® Fuse Puller
    12.7 cm (5")
    Log in for availability
    Log in for availability
    Each
    Each - 15,67 $
    15,67 $
    Each
       15,67 $
  • CA12222-726
    Large Safe-T-Grip® Fuse Puller
    Large Safe-T-Grip® Fuse Puller
    19 cm (7.5")
    Log in for availability
    Log in for availability
    Each
    Each - 23,94 $
    23,94 $
    Each
       23,94 $