ContactUsLinkComponent
E-Z Check® Plus GFI Circuit Tester, Brady Worldwide®
E-Z Check® Plus GFI Circuit Tester, Brady Worldwide®
Catalog # CA12222-722
Supplier:  Avantor
undefined
E-Z Check® Plus GFI Circuit Tester, Brady Worldwide®
Catalog # CA12222-722
Supplier:  Avantor
Specifications
  • Color:
    Yellow
  • Description:
    E-Z Check® Plus GFI Circuit Tester
  • Cat. no.:
    CA12222-722
  • Supplier No.:
    65273
Specifications
About this item
This circuit tester checks for the following conditions: correct wiring, open ground, reverse polarity, open hot, open neutral, hot on neutral, hot and ground reversed with open hot, and ground fault interruption.