Order Entry
Canada
ContactUsLinkComponent
iScript™ Select cDNA Synthesis Kit, Bio-Rad
iScript™ Select cDNA Synthesis Kit, Bio-Rad
Catalog # 101645-416
Supplier:  Bio-Rad
CAS Number:  
undefined
iScript™ Select cDNA Synthesis Kit, Bio-Rad
Catalog # 101645-416
Supplier:  Bio-Rad
Supplier Number:  1708897BUN
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    iScript™ Select cDNA Synthesis Kit
  • Cat. no.:
    101645-416
  • No. of Reactions:
    500× 20 µl
  • Supplier No.:
    1708897BUN

Specifications

About this item

The iScript™ cDNA Synthesis Kit is a sensitive and easy-to-use first-strand cDNA synthesis kit for gene expression analysis using RT-qPCR or conventional RT-PCR

  • Dilute RNA samples
  • Format allows up to 15 µl of input RNA in a 20 µL reaction
  • Low-level genes and RNA conservation
  • Broad linear dynamic range of total input RNA (1 µg–1 pg)
  • RNase A inhibitor for RNA protection during setup and reaction
  • Blend of oligo(dT) and random primers
  • Prevent 5' and 3' bias, and allow primers designed for any region

This two-tube kit is optimized for reproducible, unbiased representation over a broad dynamic range, with a fast 40 min protocol.

Ordering information: Each kit contains 5x reaction mix, iScript™ Reverse Transcriptase (RNase H+ MMLV), and nuclease-free water. Guaranteed for 12 months at —20°C in a constant temperature freezer (multiple freeze thaws not recommended).