About this item
iQ™ SYBR® Green Supermix is a 2x mix for real-time PCR (qPCR) applications
- Antibody-mediated
- Hot-start iTaq™ DNA polymerase minimizes amplification of nonspecific products
- Detection of as few as 10 copies of template
- Linear detection over 6 orders of magnitude
- Only template and primers are needed
This supermix, containing the DNA-binding dye, SYBR® Green I, is for use with sequence-specific primers, and does not require probe design. Fluorescein is included as a passive reference dye for real-time PCR detection systems that use this dye for well-factor collection.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Application:qPCR dye