Mononuclear Cell Medium, PromoCell
Consists of Basal Medium and SupplementMix, 500 ml
Catalog # 10172-392
Supplier: Promocell
Supplier #: C-28030
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The PromoCell Mononuclear Cell Medium is a very rich short-term maintenance medium which provides utmost cell viability
Recommended for Human Mononuclear Cells from Cord Blood (hMNC-CB), Human Mononuclear Cells from Peripheral Blood (hMNC-PB), CD14+ Monocytes
It is suitable to maintain MNCs for up to 48 hours before proceeding with your experiments.
Ordering information: The Medium consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Medium.
Documents
PromoCell Human Cell Culture Portfolio
Product Literature
Specifications
Specifications
- Description:Consists of Basal Medium and SupplementMix
- Size:500 ml