ContactUsLinkComponent
Ear Plug Tray, Brady®
Ear Plug Tray, Brady®
Catalog # CA82004-274
Supplier:  Brady Worldwide
undefined
Ear Plug Tray, Brady®
Catalog # CA82004-274
Supplier:  Brady Worldwide
Supplier Number:  PD228E
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Dispenser type:
    Bench top/wall-mountable
  • Material:
    Acrylic
  • No. of compartments:
    1
  • PPE type:
    Earplugs
  • Color:
    Clear
  • Description:
    Earplug tray, acrylic
  • Cat. no.:
    CA82004-274
  • W×D×H:
    10.6×10.6×10.6"
  • Supplier No.:
    PD228E
Specifications
About this item
Clear acrylic ear plug dispenser tray holds a standard box of ear plugs and the front section catches spills and protects the corrugated dispenser
Use on table top or wall mount.