You searched for: Histology Stains
Used for biological or medical research, histology stains and reagents support easy microscopic anatomy analysis. Powder or liquid form stains effectively highlight plant or animal cell and tissue components. When used in conjunction with microscopic equipment, the premium and environmentally friendly chemicals provide fast and clear observation results. From the general purpose staining combination of hematoxylin and eosin to complex kits, detect and contrast cellular sections through use of stable stains.
Stain Kits and Replacement Components, Epredia
Supplier: Epredia
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
Expand 55 Items
Alcian Blue/PAS Special Stain Kit, Epredia™
Supplier: Thermo Scientific
Demonstrate the presence of both neutral and acidic mucosubstances with this stain kit.
Expand 1 Items
Tissue Marking Dyes, Azer Scientific
Supplier: Azer Scientific
Mark critical margins and areas of interest on tissue sections with these assorted colors of tissue marking dye.
Expand 11 Items
Prussian Blue Iron Stain Kit (Reaction for Demonstration of Iron), Polysciences
Supplier: Polysciences Inc
Prussian Blue or Perls’ reaction is used to demonstrate ferric iron and ferritin
Expand 1 Items
Alcian Blue Histology Stain Kit, pH 2.5, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains acid mucins, carboxylated and sulfated mucosubstances (except strongly sulfated mucosubstances) in paraffin tissue sections
Expand 1 Items
Tissue Marking Dye Kit, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
The Original 7 Dye Color Kit is used for defining tissue margins and multiple biopsies.
Expand 4 Items
Steiner Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains fungi, Helicobacter pylori, Legionella pneumophila and Spirochetes in paraffin tissue sections
Expand 1 Items
Fungi-Fluor® Kit for Fungal Detection, Polysciences
Supplier: Polysciences Inc
The Fungi-Fluor® Kit for Fungal Detection offers a quick fluorescent stain/counterstain procedure for various fungal organisms
Expand 1 Items
P.T.A.H. Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains collagen, fibrin and striated muscle in paraffin tissue sections
Expand 1 Items
TISSUE MARKER/DYE MPC HARMONY RED 2OZ
Supplier: Mortech Manufacturing
Mopec’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in nine colors, with a standardized viscosity over the range. This ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color.
Expand 18 Items
Tissue Dye, Brush Tip, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
Brush tip tissue dye are designed for small biopsy use.
Expand 10 Items
CDI's Ink Aid, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
CDI's Ink Aid is an acetic acid solution used with tissue dye formula.
Expand 1 Items
Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.
Expand 1 Items
cAMP and cGMP conjugates, Biotium
Supplier: Biotium
Conjugates for probing cAMP receptors or measuring cGMP in immunoassays.
Expand 6 Items
Copper Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
The Copper Stain Kit (For Microwave) is intended for the demonstration of copper deposits in tissue sections.
Expand 1 Items
Tissue Marking Dye Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Fast drying, easy-to-use dye that is ideal for marking frozen or fixed tissues.
Expand 4 Items
Combined Hematoxylin and Eosin/Methenamine Silver Stain for the Histological Diagnosis of Fungi in Tissue, Electron Microscopy Sciences
Supplier: Electron Microscopy Science
A Combined Hematoxylin and Eosin/Methenamine Silver Stain is used for the Histological Diagnosis of Fungi in Tissue Sections.
Expand 1 Items
Fungi-Fluor® Pneumocystis Kit, Polysciences
Supplier: Polysciences Inc
The Fungi Fluor® Pneumocystis Kit offers a fast, fluorescent staining procedure for Pneumocystis jiroveci in bronchial specimens.
Expand 1 Items
GMS Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains fungi and Pneumocystis carinii in paraffin tissue sections
Expand 1 Items
Alcian Blue Histology Stain Kit, pH 1.0, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains strongly sulfated mucosubstances
Expand 1 Items
Fluoro-Jade B, dry powder
Supplier: Biosensis
Fluoro-Jade B stains all degenerating neurons regardless of specific insult or mechanism of cell death. The dye is highly resistant to fading and is compatible with virtually all histological processing and staining protocols.
Expand 1 Items
Mucicarmine Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains epithelial mucins in paraffin tissue sections
Expand 1 Items
Pneumocystis Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains Pneumocystis carinii in cytology smears as well as paraffin tissue sections
Expand 1 Items
Warthin-Starry Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains Helicobacter pylori, Spirochetes, Legionella pneumophila and cat scratch fever bacteria in paraffin tissue sections
Expand 1 Items
Fluoro-Jade C, dry powder
Supplier: Biosensis
Fluoro-Jade C stains all degenerating neurons regardless of specific insult or mechanism of cell death. The dye is highly resistant to fading and is compatible with virtually all histological processing and staining protocols.
Expand 1 Items
Without stand
Supplier: Mortech Manufacturing
Mopec’s range of Harmony tissue marking dyes are used to permanently mark surgical tissue margins in Histological specimens. Marking dyes are available in nine colors, with a standardized viscosity over the range. This ensures that there is less variation in viscosity from one color to another as compared to competitor products and allows the easy marking of both small and large specimens with any chosen color.
Expand 3 Items
Amyloid Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains amyloid deposits in paraffin tissue sections
Expand 1 Items
Reticulum Stain, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains reticulum fibers in paraffin tissue sections
Expand 1 Items
Von Kossa Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains calcium deposits in paraffin tissue sections
Expand 1 Items
Oil Red O Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
This kit stains fat cells and neutral fat in frozen tissue sections