About this item
The UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10208-440
- 10208-438
- Antigen name:
- CD5 molecule
- CD5 molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- UCHT2
- UCHT2
- Host:
- Mouse
- Mouse
- Gene ID:
- 921
- 921
- Isotype:
- IgG1 kappa
- IgG1 kappa