To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Erb-b2 receptor tyrosine kinase 2
- Clonality:Monoclonal
- Clone:SP3
- Gene ID:2064
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ERBB2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Cross adsorption:No
- Format:Cell Culture Supernatant
- Form:Liquid (sterile filtered)
- Antigen synonyms:NEU,TKR1,MLN 19,CD340,HER2,HER-2/neu,HER-2,NGL
- Storage buffer:0.1% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:2-8°C
- Shipping temperature:Wet Ice
- Immunogen:erbB2 monoclonal antibody was produced by repeated immunizations with a recombinant Protein corresponding to amino acid residues encoding the extracellular domain of human c-erbB2 Protein. The hybridoma was produced by fusing New Zealand White rabbit sple
- Cat. no.:RL900-C01-B37
- Supplier No.:900-C01-B37
Specifications
About this item
erbB2 Antibody monoclonal antibody contains TBS/1% BSA and is suitable for ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 185 kDa in size.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:200-1:500; Immunochemistry: 1:100
Do not freeze!
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: SP3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human