Order Entry
ContactUsLinkComponent
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (redFluor® 710)
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (redFluor® 710)
Catalog #: 10208-542
Supplier:  Tonbo Biosciences
REDFL 710 MS IGG2A ISO CTL(C1.18.4)100U
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (redFluor® 710)
Catalog #: 10208-542
Supplier:  Tonbo Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    C1.18.4
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Conjugation:
    redFluor® 710
  • Flow cytometry:
    Yes
  • Size:
    100 µg
  • Storage temperature:
    2-8°C protected from light
  • Tested applications:
    Flow Cytometry
  • Cat. no.:
    10208-542
  • Supplier No.:
    80-4724-U100

Specifications

About this item

The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: C1.18.4
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0

Customers Who Bought This Also Bought