Order Entry
Canada
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody (violetFluor® 450) [clone: 61D3]
Anti-CD14 Mouse Monoclonal Antibody (violetFluor® 450) [clone: 61D3]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD14 Mouse Monoclonal Antibody (violetFluor® 450) [clone: 61D3]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10208-502
  • 10208-500
  • Antigen symbol:
  • CD14
  • CD14
  • Antigen name:
  • CD14 Molecule
  • CD14 Molecule
  • Antigen synonyms:
  • monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
  • monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • VioletFluor® 450
  • VioletFluor® 450
  • Clone:
  • 61D3
  • 61D3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 929
  • 929
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together