To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 10
- Clonality:Monoclonal
- Clone:JES5-2A5
- Gene ID:16153
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:IL10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Size:100 µg
- Western blot:Yes
- Antigen synonyms:IL10A,IL-10,GVHDS,TGIF,CSIF
- Storage temperature:2-8°C
- Tested applications:ELISA Capture Antibody
- Cat. no.:10208-744
- Supplier No.:70-7102-U100
Specifications
About this item
The JES5-2A5 antibody is specific for mouse IL-10. IL-10 has numerous biological activities including the ability to inhibit both the synthesis of Th1 cytokines and proliferation of T cells. In the mouse, IL-10 is produced by phagocytic cells, conventional dendritic, cells, T cells, B cells, and NK cells. The JES5-2A5 antibody is a neutralizing antibody.
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES5-2A5
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse