Sub Surface Sampler, Grab Sampler® I, DWK Life Sciences
Sub Surface Grab Sampler®, 1000 ml
: 990250
Ideal for sampling from spillways, docks and other sub surfaces.
- Lightweight construction
- Eliminates surface contamination
- No need to physically enter the sample area
- Helps meet EPA requirements for water and wastewater sampling
The basic unit features a 183 cm aluminum body with clamp for securing a 1000 ml sample bottle. An attached extension allows the bottle to be opened and closed under water for discrete sampling.
- Capacity:1000 ml
- Description:Sub Surface Grab Sampler®