Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP4-25D2]
The MP4-25D2 antibody is specific for human IL-4, a pleiotropic cytokine that Th2 cell differentiation, B cell proliferation and differentiation, and B cell class switching to IgE. MP4-25D2 is a neutralizing antibody and is reported to cross react with rhesus monkey IL-4.
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human, Rhesus
- Catalog No:
- 10210-124
- 10210-122
- Antigen symbol:
- IL4
- IL4
- Antigen name:
- Interleukin 4
- Interleukin 4
- Antigen synonyms:
- BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
- BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- MP4-25D2
- MP4-25D2
- Reactivity:
- Rhesus monkey, Human
- Rhesus monkey, Human
- Host:
- Rat
- Rat
- Gene ID:
- 3565
- 3565
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- 2-8°C protected from light
- 2-8°C protected from light
- Tested applications:
- Flow Cytometry
- Flow Cytometry
Frequently Bought Together