Glass Filled Polypropylene Screw Cap, Wheaton
Catalog # 80076-570
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Description:Glass Filled PP Screw Cap with Bonded PTFE / Silicone Septa
- Cat. no.:80076-570
- Thread Size:24-400
- Supplier No.:W224600
Specifications
About this item
Glass filled Polypropylene screw cap is ideal for EPA samples
Thes vials provides greater heat resistance than normal PP caps. The cap has PTFE / silicone septa bonded to it.