Raw Fibre Extractors, VELP Scientifica
Supplier: Avantor
Certificates
About this item
Raw fibre determination is useful for nutritional, economic and legislative aspects in the food industry, where fibre content is often requirement for labelling of packaged foods.
- Rapid analysis, reliable results and high reproducibility
- Performs single or sequential extraction including boiling, rinsing and filtration
- Sample removal with air pump
- Reagent discharge with peristaltic pump
The FIWE units provide a semi automatic process to meet requirements of ADF, NDF, Weende and Van Soest methodologies.
Ordering information: FIWE units supplied with heat shield, two- place hot plate, type RC2, reagent glass bottles, glass crucibles (three or six), holder for crucibles, crucible tongs, inlet tube and PVC tubing.
Frequently Bought Together