Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Homogenizers
Tissue Grinders
Accessories for Micro Tissue Grinder with Cap, DWK Life Sciences
Accessories for Micro Tissue Grinder with Cap, DWK Life Sciences
Accessories for Micro Tissue Grinder with Cap, DWK Life Sciences
Catalog # 14231-350
Supplier:
DWK Life Sciences (KIMBLE)
Small Business Enterprise
Expand image
Accessories for Micro Tissue Grinder with Cap, DWK Life Sciences
Catalog # 14231-350
Supplier:
DWK Life Sciences (KIMBLE)
Supplier Number:
357537
Small Business Enterprise
Specifications
Description:
Replacement Pestle
Cat. no.:
14231-350
Supplier No.:
357537
Specifications
About this item
Product Details
Product Details