Model 42570 Dual Laser InfraRed Thermometer, Extech
Supplier: Avantor
Certificates
About this item
Fast response (100mS) with highest accuracy at point where dual lasers converge plus Type K input and USB interface
High 50:1 distance to target ratio measures smaller surface areas at greater distances. The 42570 features tripod mounting feature and Lock function for continuous measurements. Stores up to 100 readings and has MAX/MIN/AVG/DIF functions. USB PC interface.
Certifications: CE.
Ordering information: Complete with Type K Temperature probe, USB cable and software, tripod, carrying case and 9V battery. Three-year warranty.
Frequently Bought Together