Micro-Tissue Grinder Kit, Wheaton
Complete kit
: 358204
Micro tissue grinder kit is ideal for sterile homogenization processes.
- Autoclavable
- Replacement parts available separately
- Made from 33 low extractable borosilcate glass
The micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
Delivery information: It is supplied with 7 micro tissue grinders and the kit is packed in a high density polyethylene case with foam inserts.
- Description:Complete kit