Order Entry
Canada
ContactUsLinkComponent
Cobalt, plasma standard solution, Specpure®, Co 10 µg/ml, Thermo Scientific
Cobalt, plasma standard solution, Specpure®, Co 10 µg/ml, Thermo Scientific
Catalog # AA45252-AE
CAS Number:  
Cobalt, plasma standard solution, Specpure®, Co 10 µg/ml, Thermo Scientific
Catalog # AA45252-AE
Supplier Number:  045252-AE
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃
  • Size:
    100 ml
  • MDL No.:
    MFCD00010935
  • Cat. no.:
    AA45252-AE
  • Element:
    Cobalt
  • Supplier No.:
    45252-AE

Specifications

About this item

Cobalt, plasma standard solution, Specpure|r, Co 10µg/ml

Molecular Formula: Co in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010935