Order Entry
Canada
ContactUsLinkComponent
 
Ecosense pH10A Handheld pH/Temperature Pen Tester, YSI
  10032-016
 :  YSI
undefined
Ecosense pH10A Handheld pH/Temperature Pen Tester, YSI
  10032-016
 :  YSI
 :  PH10

 

  • Calibration:
    at 0.0 to 60 °C (32.0 to 140.0 °F)
  • Memory:
    Non-volatile; 50 Sets (pH, temperature, date and time stamp); erase all data function
  • Temperature range:
    0.0 to 100.0 °C (32.0 to 212 °F)
  • Description:
    Ecosense pH10A handheld pH/temperature pen tester
  • Operating temperature range:
    0.0…50.0 °C (32.0…122.0 °F)
  • Temperature accuracy:
    ±0.3 °C (±0.6 °F)
  • Temperature resolution:
    0.1 °C (0.2 °F)
  • Battery life:
    200 hours or greater
  • Automatic buffer recognition:
    USA (4.01, 7.00, & 10.01) or NIST (4.01, 6.86, & 9.18)
  • Temperature compensation:
    Auto 0.0 to 100.0 °C (32.0 to 212 °F)
  • Weight:
    105 g (3.7 oz.)
  • pH accuracy:
    ±0.1 unit within 10 °C of calibration, ±0.2 unit within 20 °C
  • pH range:
    0.00 to 14.00
  • pH resolution:
    0.01
  • Battery type:
    Four LR44 Alkalines
  • Cat. no.:
    10032-016
  • Supplier No.:
    PH10

 

 

The pH10A is part of the EcoSense® family of products. It's the perfect instrument for economical spot sampling of pH and temperature in many applications including wastewater, surface water, aquaculture, hydroponics, pools and education.

  • Waterproof IP 67
  • Automatic calibration and buffer recognition
  • 50-set memory (pH, temp, date and time stamp)
  • Replaceable single- or double-junction electrode
  • GLP functionality
  • 1-, 2- or 3-point calibration
  • Hold feature locks stable readings on the display
  • >200 h battery life with low battery indicator
  • Probe: Thermistor, 10kΩ / 25 °C
 : CE compliance. IP-67 Waterproof Rating.
 : Instrument includes pH electrode, batteries and instruction manual. Replacement electrodes and batteries are sold seperately. Instrument comes with a 1-year instrument and 6-month electrode warranty. The pH10A (gray) directly replaces the pH10 (blue).