Orders LinkContactUsLinkComponent
Venous Access Device Model
Venous Access Device Model
Venous Access Device Model
Catalog # 470119-452
Supplier #:  1005563
Venous Access Device Model
Catalog # 470119-452
Venous Access Device Model
Venous Access Device Model
Venous Access Device Model
Price
2 040,25 $
Log in for availability
Each
Product Details & Documents
Inexpensive Catheter Cleaning Training
  • Practice cleaning 2 catheter systems
  • Viewing port to access one of the implanted devices
  • Includes carrying case
Teach patients and healthcare staff the care and use of two different types of catheters with this model. Features left and right subclavian catheter connections that may be flushed with water; implanted venous access system with a "skin" flap over the implant to allow viewing. Made of skinned, lightweight urethane foam, this model may be washed with soap and water, or painted with antiseptics.
Mounted on a collapsible easel and packed in its own carrying case.
Specifications
  • Description:
    Venous Access Device Model