- Temperature range:−60...230 °C (−76...446 °F)
- Description:Heidolph Radleys reactor-ready pilot core lab reactor
- Cat. no.:CA10145-670
- Supplier No.:015908000CA
The Radleys reactor-ready pilot lab reactor is designed as a universal reactor workstation that can be used for different vessel sizes and different projects
Rapid vessel exchange with quick-release vessel clamp and wide-bore hose couplings. System accommodates a range of single and vacuum jacketed vessels, from 5 to 20 L. To mimic plant scale reactors, vessel manufactures at a 1.25 to 1 ratio of internal height to diameter.
The system accepts all leading brands of overhead stirrer and allows easy, tool-free adjustment. Compact stainless steel framework accepts all vessel sizes.
The self-aligning stirrer coupling engages without the need for tools. Innovative hose manifolds allow easy thermo fluid drain down. A wide range of accessory glassware is available separately.
The Radleys reactor-ready Pilot lab reactor is ideal for process development, scale-up, pilot, and kilo labs.