Orders LinkContactUsLinkComponent
Basic Buddy® Budget CPR Torsos
Supplier:   Nasco
Basic Buddy® Budget CPR Torsos
Supplier:  Nasco
Certificates
Basic Buddy® Budget CPR Torsos
Basic Buddy® Budget CPR Torsos
5 Options Available Below
Product Details & Documents
Buy Just Enough For Your Needs.
  • Includes replaceable lung bags
  • Mouth protection is included on lung bags for sanitary use
  • Available as individual, or bundles of trainers
Inexpensive manikins designed for teaching individuals or large groups the life-saving techniques of CPR. The one-piece, disposable lung/mouth protection bags make this manikin completely sanitary. The airway opens using the head tilt/chin lift method and there is a visible chest rise when ventilated. The xiphoid process provides an anatomical reference point for hand placement and compressions. The manikin also features both adult and child capabilities.
Each trainer comes with 10 lung mouth protection bags. Bundles are also supplied with 1 carrying / storage bag for every 5 Units.
Product Family Options
Simulators
Product Information
  • Availability
    Price
  • 470029-890
    Budget CPR Torso
    Budget CPR Torso
    Log in for availability
    Log in for availability
    Each
    Each - 294,20 $
    294,20 $
    Each
       294,20 $
    Supplier Number: LF03693
  • 470029-892
    Budget CPR Torso 5-Pack
    Budget CPR Torso 5-Pack
    Log in for availability
    Log in for availability
    Each
    Each - 1 324,85 $
    1 324,85 $
    Each
       1 324,85 $
    Supplier Number: LF03694
  • 470119-820
    Budget CPR Torso 10-Pack
    Budget CPR Torso 10-Pack
    Log in for availability
    Log in for availability
    Each
    Each - 2 327,40 $
    2 327,40 $
    Each
       2 327,40 $
    Supplier Number: LF03695
Replacement Materials
Product Information
  • Availability
    Price
  • 470119-822
    Replacement Lung Bags
    Replacement Lung Bags
    Log in for availability
    Log in for availability
    Each
    Each - 156,40 $
    156,40 $
    Each
       156,40 $
    Supplier Number: LF03696
  • 470119-824
    Carrying Bag
    Carrying Bag
    Log in for availability
    Log in for availability
    Each
    Each - 114,10 $
    114,10 $
    Each
       114,10 $
    Supplier Number: LF03697