Specifications
- Description:Miriad Rapid HBc/HIV/HCV Antibody Test
- Cat. no.:CA10171-070
- Supplier No.:815311005985
Specifications
About this item
Miriad Rapid HBc/HIV/HCV Antibody Test, Point-of-Use Format employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis B core antigen (anti-HBc total), HIV-1/2, and Hepatitis C
Miriad HBc/HIV/HCV features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
Universal Buffer Accessory used with Miriad Rapid HBc/HIV/HCV Antibody Test LAB+ format simplifies Point-of-Use applications. Miriad HBc/HIV/HCV employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis B core antigen (anti-HBc total), HIV-1/2, and Hepatitis C. In three easy steps users can obtain multiple instant results on a single test cartridge.
In three easy steps users can obtain multiple instant results on a single test cartridge.