hema-screen™ SPECIFIC, Fecal Immunochemical Occult Blood Test, Immunostics
10147-040
:
- Description:hema-screen™ SPECIFIC Hemoglobin Controls
- Cat. no.:10147-040
- Contents:1 Positive Control, 1 Negative Control and 1 Package Insert
- Supplier No.:HSSPCON
hema-screen SPECIFIC is a rapid fecal immunochemical test (FIT)
This test delivers a convenient, qualitative immunoassay for the determination of human hemoglobin in feces. It is a vital tool in the diagnosis and therapy of gastrointestinal disorders. hema-screen SPECIFIC features an innovative sampling method that utilizes one card for the collection of two specimens of feces and only one immunochemical specimen preparation tube or test cassette system for analysis.
: The hema-screen SPECIFIC Kit comes with 25 cassettes, 25 buffer tubes, 35 mailing envelopes, 1 product insert and 1 procedure card. The mailing envelopes contain 1 DEVEL-A-TAB slide, 2 collection tissues, 2 applicator sticks and patient instructions.