Anti-S1 tag Mouse Monoclonal Antibody [clone: 1A9B5]
Catalog # 10094-068
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:S1 tag
- Antigen symbol:S1 tag
- Clonality:Monoclonal
- Clone:1A9B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Protein A Purification
- Cat. no.:10094-068
- Supplier No.:66165-1-IG
Specifications
About this item
S1 tag, with sequence NANNPDWDF, is derived from the hepatitis B virus preS1 region. S1 tag is widely used in various protein expression systems. This antibody can detect the over-expressed fusion proteins containing S1 epitope tag.
Western Blot: Recombinant Protein, 1:5000-1:50000
Type: Primary
Antigen: S1 tag
Clonality: Monoclonal
Clone: 1A9B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human