Anti-STMN3 Mouse Monoclonal Antibody [clone: 1F12B1]
Catalog # 10095-330
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Stathmin 3
- Antigen symbol:STMN3
- Clonality:Monoclonal
- Clone:1F12B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:Sclip|SCG10-like protein|STMN3_HUMAN.|RP23-83A14.1|Scgn10 like protein
- Storage buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein G Purification
- Cat. no.:10095-330
- Supplier No.:66152-1-IG
Specifications
About this item
Western Blot: Human Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: STMN3
Clonality: Monoclonal
Clone: 1F12B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human