Anti-MBIP Mouse Monoclonal Antibody [clone: 2E6H11]
Catalog # 10090-012
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:MAP3K12-binding inhibitory protein 1
- Antigen symbol:MBIP
- Clonality:Monoclonal
- Clone:2E6H11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein A Purification
- Cat. no.:10090-012
- Supplier No.:66102-1-IG
Specifications
About this item
Western Blot:293 Cells, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200
Type: Primary
Antigen: MBIP
Clonality: Monoclonal
Clone: 2E6H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human