Anti-GMF-beta Mouse Monoclonal Antibody
Catalog # 10087-660
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Glia Maturation Factor, Beta
- Antigen symbol:GMFB
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:GMF. GMF-beta|Glia maturation factor beta|GMFB_HUMAN.|GMFB
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic acid/ammonium sulfate precipitation
- Cat. no.:10087-660
- Supplier No.:60062-1-IG
Specifications
About this item
GMFB, Glia maturation factor beta, is a nerve growth factor implicated in nervous system development. It was reported that GMF-beta promotes the phenotypic expression of glia and neurons and inhibits the proliferation of their respective tumors in culture cells. GMF-beta protein is specific to the brain where it is expressed in glial cells (mainly astrocytes) and insome neurons. Higher levels of GMFB were found in the central nervous system, except for the spinal cord, and in thymus and colon.
Western Blot: Human Heart, 1:200-1:2000; IF: MCF-7 Cells, 1:10-1:100
Type: Primary
Antigen: GMF-beta
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse