Anti-BCL11B Rabbit Polyclonal Antibody
Catalog # 10083-548
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:B-cell lymphoma/leukemia 11B
- Antigen symbol:BCL11B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10083-548
- Supplier No.:55414-1-AP
Specifications
About this item
BCL11B, also named as CTIP2, RIT1, CTIP-2, ZNF856B and hRIT1-alpha, is a tumor-suppressor protein involved in T-cell lymphomas. It may function on the P53-signaling pathway. BCL11B is a key regulator of both differentiation and survival during thymocyte development. It tepress transcription through direct, TFCOUP2-independent binding to a GC-rich response element. This antibody is specific to BCL11B.
Western Blot:Jurkat Cells, 1:200-1:1000
Type: Primary
Antigen: BCL11B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse