Anti-SGLT3/SLC5A4 Rabbit Polyclonal Antibody
10094-500
- Antibody type:Primary
- Antigen symbol:SGLT3/SLC5A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-500
- Supplier No.:24327-1-AP
Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200
Type: Primary
Antigen: SGLT3/SLC5A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat