Order Entry
Canada
ContactUsLinkComponent
Anti-LGALS3BP Rabbit Polyclonal Antibody
Anti-LGALS3BP Rabbit Polyclonal Antibody
  10089-608
Anti-LGALS3BP Rabbit Polyclonal Antibody
  10089-608
 :  23307-1-AP-150UL

 

  • Antibody type:
    Primary
  • Antigen name:
    Lectin, Galactoside-binding, Soluble, 3 binding Protein
  • Antigen symbol:
    LGALS3BP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    MAC-2-BP|BTBD17B|gp90|M2BP|CyCAP|TANGO10B|90K
  • Storage buffer:
    PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen Affinity Purified
  • Cat. no.:
    10089-608
  • Supplier No.:
    23307-1-AP

 

 

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP is a widely expressed, secreted galectin. It has been found elevated in the serum of patients with various types of breast, lung, colorectal, ovary, and endometrial cancer, and in those infected by the human immunodeficiency virus (HIV). High expression levels of LGALS3BP are associated with a shorter survival, the occurrence of metastasis or a reduced response to chemotherapy. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1. It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Immunoblotting showed that LGALS3BP is likely contained in breast milk, serum, semen, saliva, urine, and tears.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: LGALS3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse