Anti-IRGQ Rabbit Polyclonal Antibody
10088-982
- Antibody type:Primary
- Antigen name:IRGQ
- Antigen symbol:IRGQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10088-982
- Supplier No.:23239-1-AP
Western Blot:A2780 Cells, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200
Type: Primary
Antigen: IRGQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human