Anti-SORL1 Rabbit Polyclonal Antibody
10095-008
- Antibody type:Primary
- Antigen name:Sortilin-related receptor, L(DLR class) A repeats Containing
- Antigen symbol:SORL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:150 µL
- Antigen synonyms:C11orf32|SORLA|LRP9|SorLA-1|gp250|LR11
- Storage buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. no.:10095-008
- Supplier No.:22592-1-AP
IHC: Human Brain Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: SORL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat