Anti-DAND5 Rabbit Polyclonal Antibody
10085-652
- Antibody type:Primary
- Antigen name:DAN Domain family member 5, BMP antagonist
- Antigen symbol:DAND5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:CRL2|CERL2|COCO|CER2|GREM3|DANTE|SP1|CKTSF1B3
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-652
- Supplier No.:22137-1-AP
Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200
Type: Primary
Antigen: DAND5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human