Anti-CD37 Rabbit Polyclonal Antibody
10084-342
- Antibody type:Primary
- Antigen name:CD37 molecule
- Antigen symbol:CD37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:GP52-40|TSPAN26|leukocyte antigen CD37|tetraspanin-26|leukocyte surface antigen CD37|cell differentiation antigen 37|tspan-26|CD37 antigen
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-342
- Supplier No.:21044-1-AP
Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human tonsillitis, 1:20-1:200
Type: Primary
Antigen: CD37
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: