Anti-C3orf20 Rabbit Polyclonal Antibody
Catalog # 10083-854
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen symbol:C3orf20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10083-854
- Supplier No.:20906-1-AP
Specifications
About this item
Western Blot: Human Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: C3orf20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse