Anti-ZNF622 Rabbit Polyclonal Antibody
10097-366
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 622
- Antigen symbol:ZNF622
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10097-366
- Supplier No.:20804-1-AP
Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human hepatocirrhosis, 1:20-1:200; IF: Hela, 1:10-1:100
Type: Primary
Antigen: ZNF622
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat