Anti-WDR32 Rabbit Polyclonal Antibody
Catalog # 10097-102
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:WD Repeat Domain 32
- Antigen symbol:WDR32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10097-102
- Supplier No.:20483-1-AP
Specifications
About this item
WDR32, containing seven WD repeats, is also termed DDB1- and CUL4-associated factor 10 (DCAF10), which may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex . WDR32 is able to interact with DDB1 . WDR32 is supposed to be implicated in process of ubiquitin-dependent protein degradation system. Two isoforms are described and produced by alternative splicing. Proteintech’s WDR32 antibody 20483-1-AP majorly detects WDR32 isoform1 with a theoretical MW of 61 kDa.
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200; IP: HepG2 Cells, 1:200-1:1000
Type: Primary
Antigen: WDR32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat