Anti-DPH1 Rabbit Polyclonal Antibody
10091-642
- Antibody type:Primary
- Antigen name:Diphthamide biosynthesis protein 1
- Antigen symbol:DPH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:DEDSSH|DPH2L1|OVCA1|DPH2L
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10091-642
- Supplier No.:20112-1-AP
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: DPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat