Anti-MUC16 Rabbit Polyclonal Antibody
10090-596
- Antibody type:Primary
- Antigen name:Mucin 16, Cell Surface Associated
- Antigen symbol:MUC16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:FLJ14303|CA125
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10090-596
- Supplier No.:20077-1-AP
MUC16, also named as CA125, is thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. It is a serum marker that is used routinely in gynecologic practice to monitor patients with ovarian cancer. Most biochemical studies have concluded that MUC16 is a high molecular mass glycoprotein, although estimates of its size range from 200 to 2000 kDa with smaller “subunits” being described by some investigators. The antibody recognizes the C-term of MUC16.
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human