Order Entry
Canada
ContactUsLinkComponent
Anti-LDHB Rabbit Polyclonal Antibody
Anti-LDHB Rabbit Polyclonal Antibody
  10089-576
undefined
Anti-LDHB Rabbit Polyclonal Antibody
  10089-576
 :  19988-1-AP-150UL

 

  • Antibody type:
    Primary
  • Antigen name:
    Lactate dehydrogenase B
  • Antigen symbol:
    LDHB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    LDH heart subunit|LDH-H|TRG-5|LDHBD|renal carcinoma antigen NY-REN-46|LDH-B|L-lactate dehydrogenase B chain|lactate dehydrogenase H chain
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10089-576
  • Supplier No.:
    19988-1-AP

 

 

LDHB, also named as NY-REN-46 and LDH-H, belongs to the LDH/MDH superfamily and LDH family. LDHB catalyzes the reaction: (S)-lactate + NAD+ = pyruvate + NADH. It is an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway.It is glycolytic enzymes and it is positively regulated by mTOR.Defects in LDHB are a cause of hereditary LDHB deficiency. The antibody has no cross reaction with LDHA and LDHC.

Western Blot: HeLa Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: LDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat