Anti-CCT6A Rabbit Polyclonal Antibody
Catalog # 10084-252
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Chaperonin Containing TCP1 Subunit 6A
- Antigen symbol:CCT6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:CCT-zeta-1|CCT6A|Acute morphine dependence-related protein 2|TCP-1-zeta|Tcp20|CCT zeta-1|TCP-1- zeta|CCT6|TCP1 zeta|HTR3|TCP 1 zeta|TCP1- zeta|CCT- zeta-1|CCT- zeta1|T-complex protein 1 subunit zeta|CCT zeta1|CCTZ
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. no.:10084-252
- Supplier No.:19793-1-AP
Specifications
About this item
CCT6A, also named as CCT6, CCTZ, CCT-zeta,Tcp20 and HTR3, belongs to the TCP-1 chaperonin family. CCT6A is a molecular chaperone which assists the folding of proteins upon ATP hydrolysis. CCT6A is known to play a role, in vitro, in the folding of actin and tubulin. The antibody has no cross reaction to CCT6B.
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: CCT6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse