Order Entry
Canada
ContactUsLinkComponent
 
Anti-HMP19 Rabbit Polyclonal Antibody
  10088-358
Anti-HMP19 Rabbit Polyclonal Antibody
  10088-358
 :  19423-1-AP-150UL

 

  • Antibody type:
    Primary
  • Antigen name:
    HMP19
  • Antigen symbol:
    HMP19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10088-358
  • Supplier No.:
    19423-1-AP

 

 

IHC: Human hepatocirrhosis, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: HMP19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat