Anti-GSR Rabbit Polyclonal Antibody
Catalog # 10087-908
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Glutathione reductase
- Antigen symbol:GSR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:HEL-75
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10087-908
- Supplier No.:18257-1-AP
Specifications
About this item
GSR(Glutathione reductase) is also named as GLUR, GRD1 and belongs to the class-I pyridine nucleotide-disulfide oxidoreductase family.It was first found in a black American a variant red cell GSR characterized by greater electrophoretic mobility and enzyme activity per unit of hemoglobin than the normal.This protein maintains high levels of reduced glutathione in the cytosol.It has 5 isoforms produced by alternative splicing and alternative initiation.It can also exsit as a homodimer with the molecular weight of 110KDa.This antibody is specific to GSR.
Western Blot:Jurkat Cells, 1:500-1:5000
Type: Primary
Antigen: GSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat